Methyl-2,6-dichloro-5-(trifluoromethyl)-4-pyrimidinecarboxylate

Lieferant: Apollo Scientific

PC200324-500MG
APOSPC200324-500MGEA 412 EUR
APOSPC200324-500MG
Methyl-2,6-dichloro-5-(trifluoromethyl)-4-pyrimidinecarboxylate
Methyl-2,6-dichloro-5-(trifluoromethyl)-4-pyrimidinecarboxylate
Formel: C₇H₃Cl₂F₃N₂O₂
Molecular Weight: 275,02 g/mol
Schmelzpunkt: 57…58 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD12922697
CAS-Nummer: 1190198-37-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR