Ethyl-8-iod-2,6-bis(trifluormethyl)imidazo[1,2-a]pyridin-2-carboxylat
Lieferant: Apollo Scientific
Synonyme:
Ethyl 2,6-bis(trifluoromethyl)-8-iodoimidazo[1,2-a]pyridine-3-carboxylate
PC200325-500MG
APOSPC200325-500MGEA
344
EUR
APOSPC200325-500MG
Ethyl-8-iod-2,6-bis(trifluormethyl)imidazo[1,2-a]pyridin-2-carboxylat
Ethyl-8-iod-2,6-bis(trifluormethyl)imidazo[1,2-a]pyridin-2-carboxylat
Formel:
C₁₂H₇F₆IN₂O₂ Schmelzpunkt: 167…168 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD12498697 CAS-Nummer: 1171918-86-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...