5-Chloro-6-cyclopropyl-2-hydroxy-4-(trifluoromethyl)nicotinonitrile

Lieferant: Apollo Scientific

PC200326-500MG
APOSPC200326-500MGEA 277 EUR
APOSPC200326-500MG
5-Chloro-6-cyclopropyl-2-hydroxy-4-(trifluoromethyl)nicotinonitrile
5-Chloro-6-cyclopropyl-2-hydroxy-4-(trifluoromethyl)nicotinonitrile
Formel: C₁₀H₆ClF₃N₂O
Schmelzpunkt: 240 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11553041
CAS-Nummer: 1092352-56-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR