3-Nitro-5-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine

Lieferant: Apollo Scientific

PC200328-500MG
APOSPC200328-500MGEA 412 EUR
APOSPC200328-500MG
3-Nitro-5-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine
3-Nitro-5-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine
Formel: C₈H₄F₃N₃O₂
Schmelzpunkt: 222…223 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11100250
CAS-Nummer: 1048914-26-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR