tert-Butyl-3-iodo-5-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine-1-carboxylate

Lieferant: Apollo Scientific

Synonyme: 3-Iodo-5-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine, N1-BOC protected, tert-Butyl 3-iodo-5-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine-1-carboxylate

PC200330-500MG
APOSPC200330-500MGEA 561 EUR
APOSPC200330-500MG
tert-Butyl-3-iodo-5-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine-1-carboxylate
tert-Butyl-3-iodo-5-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine-1-carboxylate
Formel: C₁₃H₁₂F₃IN₂O₂
Molecular Weight: 412,15 g/mol
Schmelzpunkt: 125…127 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD16622804
CAS-Nummer: 1228183-72-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR