(Morpholinium-4-ylmethyl)trifluoroborate internal salt 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC200335-1G
APOSPC200335-1GEA
98.6
EUR
APOSPC200335-1G
(Morpholinium-4-ylmethyl)trifluoroborate internal salt 95%
(Morpholinium-4-ylmethyl)trifluoroborate internal salt
Formel:
C₅H₁₁BF₃NO Molecular Weight: 168.95 g/mol Schmelzpunkt: 170…172 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD20134168 CAS-Nummer: 1268340-94-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...