4-(4-Ethyl-1H-1,2,3-triazol-1-yl)-2-fluoroaniline
Lieferant: Apollo Scientific
PC200336-500MG
APOSPC200336-500MGEA
184
EUR
APOSPC200336-500MG
4-(4-Ethyl-1H-1,2,3-triazol-1-yl)-2-fluoroaniline
4-(4-Ethyl-1H-1,2,3-triazol-1-yl)-2-fluoroaniline
Formel:
C₁₀H₁₁FN₄ Schmelzpunkt: 58…61 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD20921593 CAS-Nummer: 1276538-16-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...