4-Hydrazinyl-2,6-bis(trifluoromethyl)pyridine

Lieferant: Apollo Scientific

PC200337-500MG
APOSPC200337-500MGEA 611 EUR
APOSPC200337-500MG
4-Hydrazinyl-2,6-bis(trifluoromethyl)pyridine
4-Hydrazinyl-2,6-bis(trifluoromethyl)pyridine
Formel: C₇H₅F₆N₃
Schmelzpunkt: 124…126 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01936019
CAS-Nummer: 240115-88-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR