4-Methyl-2-[4-(trifluoromethyl)phenyl]benzoic acid

Lieferant: Apollo Scientific

PC200340-1G PC200340-500MG
APOSPC200340-1GEA 364 EUR
APOSPC200340-1G APOSPC200340-500MG
4-Methyl-2-[4-(trifluoromethyl)phenyl]benzoic acid
4-Methyl-2-[4-(trifluoromethyl)phenyl]benzoic acid
Formel: C₁₅H₁₁F₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18425410
CAS-Nummer: 273727-19-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR