Potassium pyrimidine-5-trifluoroborate

Lieferant: Apollo Scientific

PC200342-1G
APOSPC200342-1GEA 0 EUR
APOSPC200342-1G
Potassium pyrimidine-5-trifluoroborate
Potassium pyrimidine-5-trifluoroborate
Formel: C₄H₃BF₃KN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09992922
CAS-Nummer: 1242733-91-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR