N-(2-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]amino}ethyl)-2-isocyanoacetamide

Lieferant: Apollo Scientific

PC200344-500MG
APOSPC200344-500MGEA 336 EUR
APOSPC200344-500MG
N-(2-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]amino}ethyl)-2-isocyanoacetamide
N-(2-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]amino}ethyl)-2-isocyanoacetamide
Formel: C₁₁H₁₀ClF₃N₄O
Molecular Weight: 306.672 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22375447
CAS-Nummer: 1452577-23-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR