2-[3-(Trifluoromethyl)phenyl]-3,4-dihydropyrimidin-4-one

Lieferant: Apollo Scientific

PC200345-500MG
APOSPC200345-500MGEA 271 EUR
APOSPC200345-500MG
2-[3-(Trifluoromethyl)phenyl]-3,4-dihydropyrimidin-4-one
2-[3-(Trifluoromethyl)phenyl]-3,4-dihydropyrimidin-4-one
Formel: C₁₁H₇F₃N₂O
Molecular Weight: 240,181 g/mol
Schmelzpunkt: 185…187 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD12107651
CAS-Nummer: 125903-84-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR