4-{[(4-Chloro-2-fluorophenyl)amino]methyl}piperidin-4-ol

Lieferant: Apollo Scientific

PC200350-500MG
APOSPC200350-500MGEA 364 EUR
APOSPC200350-500MG
4-{[(4-Chloro-2-fluorophenyl)amino]methyl}piperidin-4-ol
4-{[(4-Chloro-2-fluorophenyl)amino]methyl}piperidin-4-ol
Formel: C₁₂H₁₆ClFN₂O
Schmelzpunkt: 131…132 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD20232914
CAS-Nummer: 1335112-98-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR