(R)-Boroleucine-(1S,2S,3R,5S)-(+)-pinanediol ester trifluoroacetate 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (R)-3-Methyl-1-((3aS,4S,6S,7aR)-3a,5,5-trimethylhexahydro-4,6-methanobenzo[d][1,3,2]dioxaborol-2-yl)butan-1-amine 2,2,2-trifluoroacetate, Bortezomib Amino R Isomer (TFA salt), (R)-BoroLeu-(+)-Pinanediol trifluoroacetate

PC200351-1G
APOSPC200351-1GEA 24.8 EUR
APOSPC200351-1G
(R)-Boroleucine-(1S,2S,3R,5S)-(+)-pinanediol ester trifluoroacetate 98%
(R)-Boroleucine-(1S,2S,3R,5S)-(+)-pinanediol ester trifluoroacetate
Formel: C₁₇H₂₉BF₃NO₄
Molecular Weight: 379.23 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD10566030
CAS-Nummer: 179324-87-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR