6-Brom-4-fluor-1H-indazol 97%

Lieferant: Apollo Scientific
Gefahr

PC200355-1G PC200355-250MG PC200355-5G
APOSPC200355-1GEA 33.5 EUR
APOSPC200355-1G APOSPC200355-250MG APOSPC200355-5G
6-Brom-4-fluor-1H-indazol 97%
6-Bromo-4-fluoro-1H-indazole
Formel: C₇H₄BrFN₂
Molecular Weight: 215.0200 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07781444
CAS-Nummer: 885520-23-0
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR