4,4-Diethoxy-1,1,1-trifluorobut-3-en-2-one
Lieferant: Apollo Scientific
PC200357-500MG
APOSPC200357-500MGEA
539
EUR
APOSPC200357-500MG
4,4-Diethoxy-1,1,1-trifluorobut-3-en-2-one
4,4-Diethoxy-1,1,1-trifluorobut-3-en-2-one
Formel:
C₈H₁₁F₃O₃ Siedepunkt: 110…122 °C/13mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD08273801 CAS-Nummer: 40657-29-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...