3-(4-Chloro-2-fluorophenyl)-1-oxa-3,8-diazaspiro[4.5]decan-2-one

Lieferant: Apollo Scientific
Achtung

PC200359-500MG
APOSPC200359-500MGEA 611 EUR
APOSPC200359-500MG
3-(4-Chloro-2-fluorophenyl)-1-oxa-3,8-diazaspiro[4.5]decan-2-one
3-(4-Chloro-2-fluorophenyl)-1-oxa-3,8-diazaspiro[4.5]decan-2-one
Formel: C₁₃H₁₄ClFN₂O₂
Schmelzpunkt: 96…97 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD20232921
CAS-Nummer: 1335220-63-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR