3-(4-Fluorophenyl)-1-oxa-3,8-diazaspiro[4.5]decan-2-one

Lieferant: Apollo Scientific

PC200360-500MG
APOSPC200360-500MGEA 611 EUR
APOSPC200360-500MG
3-(4-Fluorophenyl)-1-oxa-3,8-diazaspiro[4.5]decan-2-one
3-(4-Fluorophenyl)-1-oxa-3,8-diazaspiro[4.5]decan-2-one
Formel: C₁₃H₁₅FN₂O₂
Molecular Weight: 250,28 g/mol
Schmelzpunkt: 114…115 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD20232922
CAS-Nummer: 1047655-95-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR