Ethyl 2-(trifluoromethyl)-1H-1,3-benzodiazole-5-carboxylate

Lieferant: Apollo Scientific

PC200376-1G
APOSPC200376-1GEA 245 EUR
APOSPC200376-1G
Ethyl 2-(trifluoromethyl)-1H-1,3-benzodiazole-5-carboxylate
Ethyl 2-(trifluoromethyl)-1H-1,3-benzodiazole-5-carboxylate
Formel: C₁₁H₉F₃N₂O₂
Molecular Weight: 258,197 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01658435
CAS-Nummer: 89457-09-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR