Methyl (2R)-2-{[(tert-butoxy)carbonyl]amino}-2-{4-[(trifluoromethane)sulphonyloxy]phenyl}acetate

Lieferant: Apollo Scientific

PC200379-1G
APOSPC200379-1GEA 336 EUR
APOSPC200379-1G
Methyl (2R)-2-{[(tert-butoxy)carbonyl]amino}-2-{4-[(trifluoromethane)sulphonyloxy]phenyl}acetate
Methyl (2R)-2-{[(tert-butoxy)carbonyl]amino}-2-{4-[(trifluoromethane)sulphonyloxy]phenyl}acetate
Formel: C₁₅H₁₈F₃NO₇S
Molecular Weight: 413,366 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26523311
CAS-Nummer: 313490-19-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR