7-Chloro-6-fluoroisoquinoline

Lieferant: Apollo Scientific
Achtung

PC200380-1G
APOSPC200380-1GEA 538 EUR
APOSPC200380-1G
7-Chloro-6-fluoroisoquinoline
7-Chloro-6-fluoroisoquinoline
Formel: C₉H₅ClFN
Lagertemperatur: Raumtemperatur
MDL: MFCD11846299
CAS-Nummer: 918490-51-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR