Potassium (3-bromo-5-methoxyphenyl)trifluoroboranuide

Lieferant: Apollo Scientific

PC200382-500G
APOSPC200382-500GEA 109 EUR
APOSPC200382-500G
Potassium (3-bromo-5-methoxyphenyl)trifluoroboranuide
Potassium (3-bromo-5-methoxyphenyl)trifluoroboranuide
Formel: C₇H₆BBrF₃O·K
Molecular Weight: 292,93 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26936348
CAS-Nummer: 1189097-39-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR