Kalium(4-acetylphenyl)trifluoroborat

Lieferant: Apollo Scientific

Synonyme: Potassium 4-acetylphenyltrifluoroborate, Potassium (4-acetylphenyl)trifluoroboranuide

PC200386-5G
APOSPC200386-5GEA 63.8 EUR
APOSPC200386-5G
Kalium(4-acetylphenyl)trifluoroborat
Kalium(4-acetylphenyl)trifluoroborat
Formel: C₈H₇BF₃KO
Molecular Weight: 226.05 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07784376
CAS-Nummer: 252726-24-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR