6-Chloro-N-[3-(trifluoromethyl)phenyl]pyrimidin-4-amine

Lieferant: Apollo Scientific

PC200391-1G
APOSPC200391-1GEA 538 EUR
APOSPC200391-1G
6-Chloro-N-[3-(trifluoromethyl)phenyl]pyrimidin-4-amine
6-Chloro-N-[3-(trifluoromethyl)phenyl]pyrimidin-4-amine
Formel: C₁₁H₇ClF₃N₃
Molecular Weight: 273,642 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10000669
CAS-Nummer: 872510-82-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR