4,6-Dichloro-2-(trifluoromethyl)pyrimidine

Lieferant: Apollo Scientific

PC200402-10G PC200402-1G PC200402-5G
APOSPC200402-10GEA 613 EUR
APOSPC200402-10G APOSPC200402-1G APOSPC200402-5G
4,6-Dichloro-2-(trifluoromethyl)pyrimidine
4,6-Dichloro-2-(trifluoromethyl)pyrimidine
Formel: C₅HCl₂F₃N₂
Molecular Weight: 216.98 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08436597
CAS-Nummer: 705-24-8

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR