Methyl 2,6-difluoropyridine-3-carboxylate

Lieferant: Apollo Scientific

Synonyme: 2,6-Difluoro-3-pyridinecarboxylic acid methyl ester, Methyl 2,6-difluoronicotinate

PC200403-1G
APOSPC200403-1GEA 161 EUR
APOSPC200403-1G
Methyl 2,6-difluoropyridine-3-carboxylate
Methyl 2,6-difluoropyridine-3-carboxylate
Formel: C₇H₅F₂NO₂
Molecular Weight: 173,117 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08704426
CAS-Nummer: 117671-02-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR