Trifluoroacetohydrazide, tech
Lieferant: Apollo Scientific
|
Gefahr
|
PC200408-25G
APOSPC200408-25GEA
84.1
EUR
APOSPC200408-25G
Trifluoroacetohydrazide, tech
Trifluoroacetohydrazide
Formel:
C₂H₃F₃N₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00221440 CAS-Nummer: 1538-08-5 UN: 2923 ADR: 8+6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...