1-Methyl-4-[3-(trifluoromethyl)pyridin-2-yl]piperazine
Lieferant: Apollo Scientific
PC200413-250MG
APOSPC200413-250MGEA
217
EUR
APOSPC200413-250MG
1-Methyl-4-[3-(trifluoromethyl)pyridin-2-yl]piperazine
1-Methyl-4-[3-(trifluoromethyl)pyridin-2-yl]piperazine
Formel:
C₁₁H₁₄F₃N₃ Molecular Weight: 245.244 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22573504 CAS-Nummer: 220459-53-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...