N-Aminopyridazinium hexafluorophosphate

Lieferant: Apollo Scientific
Achtung

PC200429-1G
APOSPC200429-1GEA 270 EUR
APOSPC200429-1G
N-Aminopyridazinium hexafluorophosphate
N-Aminopyridazinium hexafluorophosphate
Formel: C₄H₆N₃·PF₆
Molecular Weight: 241,075 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27924239
CAS-Nummer: 346412-97-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR