1,3-Dibromo-2-fluoro-4-methoxybenzene

Lieferant: Apollo Scientific

PC200430-500MG
APOSPC200430-500MGEA 396 EUR
APOSPC200430-500MG
1,3-Dibromo-2-fluoro-4-methoxybenzene
1,3-Dibromo-2-fluoro-4-methoxybenzene
Formel: C₇H₅Br₂FO
Molecular Weight: 283,92 g/mol
Schmelzpunkt: 92…94 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD27957293
CAS-Nummer: 1526600-49-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR