1-[2-(4-Fluorophenyl)-4-methyl-1,3-oxazol-5-yl]ethan-1-one

Lieferant: Apollo Scientific

PC200433-1G
APOSPC200433-1GEA 396 EUR
APOSPC200433-1G
1-[2-(4-Fluorophenyl)-4-methyl-1,3-oxazol-5-yl]ethan-1-one
1-[2-(4-Fluorophenyl)-4-methyl-1,3-oxazol-5-yl]ethan-1-one
Formel: C₁₂H₁₀FNO₂
Molecular Weight: 219,212 g/mol
Schmelzpunkt: 90…92 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD27957301
CAS-Nummer: 1565845-75-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR