Ethyl 3-cyano-6-methyl-4-(trifluoromethyl)pyridine-2-carboxylate

Lieferant: Apollo Scientific

PC200434-1G
APOSPC200434-1GEA 663 EUR
APOSPC200434-1G
Ethyl 3-cyano-6-methyl-4-(trifluoromethyl)pyridine-2-carboxylate
Ethyl 3-cyano-6-methyl-4-(trifluoromethyl)pyridine-2-carboxylate
Formel: C₁₁H₉F₃N₂O₂
Schmelzpunkt: 57…59 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD27957302
CAS-Nummer: 1565827-82-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR