2-Fluor-(4-methoxycarbonyl)phenylboronsäurepinakolester

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 2-Fluoro-4-(methoxycarbonyl)phenylboronic acid,pinacol ester, 2-Fluoro-4-(methoxycarbonyl)benzeneboronic acid pinacol ester

PC200435-5G
APOSPC200435-5GEA 148 EUR
APOSPC200435-5G
2-Fluor-(4-methoxycarbonyl)phenylboronsäurepinakolester
2-Fluor-(4-methoxycarbonyl)phenylboronsäurepinakolester
Formel: C₁₄H₁₈BFO₄
Molecular Weight: 280.0997 g/mol
Schmelzpunkt: 66…71 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11855962
CAS-Nummer: 603122-79-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR