(4-Aminophenyl)(4-fluorophenyl)methanone

Lieferant: Apollo Scientific

Synonyme: 4-[(4-Fluorophenyl)carbonyl]aniline

PC200436-1G
APOSPC200436-1GEA 161 EUR
APOSPC200436-1G
(4-Aminophenyl)(4-fluorophenyl)methanone
(4-Aminophenyl)(4-fluorophenyl)methanone
Formel: C₁₃H₁₀FNO
Molecular Weight: 215,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00498555
CAS-Nummer: 10055-40-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR