Potassium trifluoro[3-(2-methoxyethoxy)phenyl]boranuide

Lieferant: Apollo Scientific

PC200437-5G
APOSPC200437-5GEA 430 EUR
APOSPC200437-5G
Potassium trifluoro[3-(2-methoxyethoxy)phenyl]boranuide
Potassium trifluoro[3-(2-methoxyethoxy)phenyl]boranuide
Formel: C₉H₁₁BF₃O₂·K
Molecular Weight: 258,087 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18861625
CAS-Nummer: 1985700-28-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR