3-Cyano-6-methyl-4-(trifluoromethyl)pyridine-2-carboxylic acid
Lieferant: Apollo Scientific
PC200444-500MG
APOSPC200444-500MGEA
661
EUR
APOSPC200444-500MG
3-Cyano-6-methyl-4-(trifluoromethyl)pyridine-2-carboxylic acid
3-Cyano-6-methyl-4-(trifluoromethyl)pyridine-2-carboxylic acid
Formel:
C₉H₅F₃N₂O₂ Molecular Weight: 230,143 g/mol Schmelzpunkt: 160 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD27957326 CAS-Nummer: 1565827-83-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...