2-Chloro-6-(trifluoromethoxy)pyridine
Lieferant: Apollo Scientific
|
Achtung
|
PC200446-1G
APOSPC200446-1GEA
209
EUR
APOSPC200446-1G
2-Chloro-6-(trifluoromethoxy)pyridine
2-Chloro-6-(trifluoromethoxy)pyridine
Formel:
C₆H₃ClF₃NO Siedepunkt: 81…83 °C (26 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD18909427 CAS-Nummer: 1221171-70-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...