6-Chloro-2-(trifluoromethoxy)nicotinic acid

Lieferant: Apollo Scientific
Achtung

PC200447-1G
APOSPC200447-1GEA 949 EUR
APOSPC200447-1G
6-Chloro-2-(trifluoromethoxy)nicotinic acid
6-Chloro-2-(trifluoromethoxy)nicotinic acid
Formel: C₇H₃ClF₃NO₃
Molecular Weight: 241,552 g/mol
Schmelzpunkt: 139…142 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD22571781
CAS-Nummer: 1221171-91-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR