1-[5-Chloro-4-(difluoromethyl)pyridin-2-yl]piperazine

Lieferant: Apollo Scientific

PC200448-1G
APOSPC200448-1GEA 529 EUR
APOSPC200448-1G
1-[5-Chloro-4-(difluoromethyl)pyridin-2-yl]piperazine
1-[5-Chloro-4-(difluoromethyl)pyridin-2-yl]piperazine
Formel: C₁₀H₁₂ClF₂N₃
Molecular Weight: 247,672 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27957335
CAS-Nummer: 1565827-81-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR