Ethyl 5-(3,4-difluorophenyl)-1-methyl-1H-pyrazole-3-carboxylate
Lieferant: Apollo Scientific
PC200449-1G
APOSPC200449-1GEA
538
EUR
APOSPC200449-1G
Ethyl 5-(3,4-difluorophenyl)-1-methyl-1H-pyrazole-3-carboxylate
Ethyl 5-(3,4-difluorophenyl)-1-methyl-1H-pyrazole-3-carboxylate
Formel:
C₁₃H₁₂F₂N₂O₂ Molecular Weight: 266.243 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14522979 CAS-Nummer: 1170363-38-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...