4-(4-Methoxyphenyl)-1-methyl-3-(trifluoromethyl)-1H-pyrazol-5-amine

Lieferant: Apollo Scientific

PC200450-1G
APOSPC200450-1GEA 538 EUR
APOSPC200450-1G
4-(4-Methoxyphenyl)-1-methyl-3-(trifluoromethyl)-1H-pyrazol-5-amine
4-(4-Methoxyphenyl)-1-methyl-3-(trifluoromethyl)-1H-pyrazol-5-amine
Formel: C₁₂H₁₂F₃N₃O
Molecular Weight: 271,238 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12653214
CAS-Nummer: 164668-12-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR