1-(2-{[3-(Trifluoromethyl)phenyl]amino}-1,3-thiazol-5-yl)ethan-1-one
Lieferant: Apollo Scientific
PC200452-1G
APOSPC200452-1GEA
408
EUR
APOSPC200452-1G
1-(2-{[3-(Trifluoromethyl)phenyl]amino}-1,3-thiazol-5-yl)ethan-1-one
1-(2-{[3-(Trifluoromethyl)phenyl]amino}-1,3-thiazol-5-yl)ethan-1-one
Formel:
C₁₂H₉F₃N₂OS Molecular Weight: 286,273 g/mol Schmelzpunkt: 193…195 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD27980587 CAS-Nummer: 1582770-06-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...