Methyl 6-oxo-1-{[4-(trifluoromethyl)phenyl]methyl}-1,6-dihydropyridine-3-carboxylate

Lieferant: Apollo Scientific

PC200453-1G
APOSPC200453-1GEA 277 EUR
APOSPC200453-1G
Methyl 6-oxo-1-{[4-(trifluoromethyl)phenyl]methyl}-1,6-dihydropyridine-3-carboxylate
Methyl 6-oxo-1-{[4-(trifluoromethyl)phenyl]methyl}-1,6-dihydropyridine-3-carboxylate
Formel: C₁₅H₁₂F₃NO₃
Schmelzpunkt: 103…105 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD27980590
CAS-Nummer: 1582770-08-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR