5-Amino-1-(2,2,2-trifluoroethyl)-1H-pyrazole-4-carbonitrile

Lieferant: Apollo Scientific

PC200455-1G
APOSPC200455-1GEA 894 EUR
APOSPC200455-1G
5-Amino-1-(2,2,2-trifluoroethyl)-1H-pyrazole-4-carbonitrile
5-Amino-1-(2,2,2-trifluoroethyl)-1H-pyrazole-4-carbonitrile
Formel: C₆H₅F₃N₄
Molecular Weight: 190,126 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00128420
CAS-Nummer: 1082745-56-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR