5-Amino-1-(2,2,2-trifluoroethyl)-3-methyl-1H-pyrazole-4-carbonitrile

Lieferant: Apollo Scientific

PC200456-1G
APOSPC200456-1GEA 684 EUR
APOSPC200456-1G
5-Amino-1-(2,2,2-trifluoroethyl)-3-methyl-1H-pyrazole-4-carbonitrile
5-Amino-1-(2,2,2-trifluoroethyl)-3-methyl-1H-pyrazole-4-carbonitrile
Formel: C₇H₇F₃N₄
Molecular Weight: 204,152 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04973473
CAS-Nummer: 168465-05-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR