2,3-Dichloro-4-(difluoromethyl)pyridine

Lieferant: Apollo Scientific

PC200457-500MG
APOSPC200457-500MGEA 438 EUR
APOSPC200457-500MG
2,3-Dichloro-4-(difluoromethyl)pyridine
2,3-Dichloro-4-(difluoromethyl)pyridine
Formel: C₆H₃Cl₂F₂N
Lagertemperatur: Raumtemperatur
MDL: MFCD22418352
CAS-Nummer: 1257071-85-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR