2,4-Dichloro-5-(difluoromethyl)pyridine

Lieferant: Apollo Scientific
Achtung

PC200458-1G
APOSPC200458-1GEA 671 EUR
APOSPC200458-1G
2,4-Dichloro-5-(difluoromethyl)pyridine
2,4-Dichloro-5-(difluoromethyl)pyridine
Formel: C₆H₃Cl₂F₂N
Molecular Weight: 197,998 g/mol
Schmelzpunkt: 37…40 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD18257012
CAS-Nummer: 1256833-30-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR