2-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-6-(trifluoromethyl)pyrimidin-4-ol

Lieferant: Apollo Scientific

PC200459-500MG
APOSPC200459-500MGEA 205 EUR
APOSPC200459-500MG
2-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-6-(trifluoromethyl)pyrimidin-4-ol
2-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-6-(trifluoromethyl)pyrimidin-4-ol
Formel: C₁₁H₄ClF₆N₃O
Molecular Weight: 343,612 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27980616
CAS-Nummer: 1582770-04-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR