Ethyl-2-cyano-3-[3-(trifluoromethoxy)phenyl]prop-2-enoate

Lieferant: Apollo Scientific

PC200461-1G
APOSPC200461-1GEA 147 EUR
APOSPC200461-1G
Ethyl-2-cyano-3-[3-(trifluoromethoxy)phenyl]prop-2-enoate
Ethyl-2-cyano-3-[3-(trifluoromethoxy)phenyl]prop-2-enoate
Formel: C₁₃H₁₀F₃NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD28023499
CAS-Nummer: 773089-44-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR