N-(2-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}ethyl)acetamide

Lieferant: Apollo Scientific

PC200462-1G
APOSPC200462-1GEA 423 EUR
APOSPC200462-1G
N-(2-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}ethyl)acetamide
N-(2-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}ethyl)acetamide
Formel: C₁₀H₁₀ClF₃N₂O₂
Molecular Weight: 282,65 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28023503
CAS-Nummer: 1610047-55-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR